Assay Details
| Gene Symbol: KT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336078.1 | NP_001324538.1 | 7 - 8 | 1502 | 80 | |
| NM_128222.6 | NP_180233.1 | 8 - 9 | 1757 | 80 | ||
| GenBank mRNA | AK317729.1 | - | 8 - 9 | 1528 | 80 | |
| BT006442.1 | - | 7 - 8 | 1148 | 80 | ||
| X62907.1 | - | 1520 | 80 | |||
More Information
Set Membership: |
|

KT1
Genomic Map