Assay Details
| Gene Symbol: NTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128289.2 | NP_180298.1 | 2 - 3 | 455 | 101 | |
| GenBank mRNA | BT028934.1 | - | 2 - 3 | 439 | 101 | |
| DQ056544.1 | - | 2 - 3 | 439 | 101 | ||
More Information
Set Membership: |
|

NTL8
Genomic Map