Assay Details
| Gene Symbol: ROPGEF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113373.2 | NP_189105.1 | 2 - 3 | 485 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ROPGEF8| Gene Symbol: ROPGEF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113373.2 | NP_189105.1 | 2 - 3 | 485 | 69 | |
Set Membership: |
|