Assay Details
Gene Symbol: ARP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113614.3 | NP_189336.1 | 7 - 8 | 787 | 84 | |
GenBank mRNA | AF507910.1 | - | 7 - 8 | 589 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
