Assay Details
| Gene Symbol: ARP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113614.3 | NP_189336.1 | 9 - 10 | 926 | 86 | |
| GenBank mRNA | AF507910.1 | - | 9 - 10 | 728 | 86 | |
More Information
Set Membership: |
|

ARP2
Genomic Map