Assay Details
| Gene Symbol: MUM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125775.4 | NP_568978.2 | 15 - 16 | 2274 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MUM2| Gene Symbol: MUM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125775.4 | NP_568978.2 | 15 - 16 | 2274 | 82 | |
Set Membership: |
|