Assay Details
Gene Symbol: BGAL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125776.2 | NP_201186.1 | 10 - 11 | 1265 | 123 | |
GenBank mRNA | AY093118.1 | - | 10 - 11 | 1265 | 123 | |
BT010338.1 | - | 10 - 11 | 1097 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
