Assay Details
Gene Symbol: BGAL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125776.2 | NP_201186.1 | 3 - 4 | 559 | 97 | |
GenBank mRNA | AY093118.1 | - | 3 - 4 | 559 | 97 | |
BT010338.1 | - | 3 - 4 | 391 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
