Assay Details
| Gene Symbol: FMO GS-OX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084053.1 | NP_001077522.1 | 3 - 4 | 778 | 66 | |
| NM_001332018.1 | NP_001323223.1 | 3 - 4 | 784 | 66 | ||
| NM_101086.5 | NP_172678.3 | 3 - 4 | 868 | 66 | ||
| GenBank mRNA | BT030387.1 | - | 3 - 4 | 767 | 66 | |
More Information
Set Membership: |
|

FMO GS-OX5
Genomic Map