Assay Details
| Gene Symbol: NHX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198066.1 | NP_001184995.1 | 10 - 11 | 1019 | 72 | |
| NM_101333.5 | NP_172918.2 | 10 - 11 | 1152 | 72 | ||
| GenBank mRNA | DQ104324.1 | - | 10 - 11 | 990 | 72 | |
More Information
Set Membership: |
|

NHX8
Genomic Map