Assay Details
| Gene Symbol: ADH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106362.3 | NP_177837.1 | 5 - 6 | 1115 | 65 | |
| GenBank mRNA | AY045612.1 | - | 5 - 6 | 916 | 65 | |
| AY090330.1 | - | 5 - 6 | 861 | 65 | ||
More Information
Set Membership: |
|

ADH1
Genomic Map