Assay Details
Gene Symbol: GGH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198500.1 | NP_001185429.1 | 6 - 7 | 921 | 127 | |
NM_106515.4 | NP_565186.2 | 6 - 7 | 896 | 127 | ||
GenBank mRNA | AY070047.1 | - | 6 - 7 | 812 | 127 | |
AY096428.1 | - | 6 - 7 | 744 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
