Assay Details
| Gene Symbol: AT1G79060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106557.4 | NP_178027.1 | 1 | 1751 | 138 | |
| GenBank mRNA | AK228755.1 | - | 1 | 1710 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G79060| Gene Symbol: AT1G79060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106557.4 | NP_178027.1 | 1 | 1751 | 138 | |
| GenBank mRNA | AK228755.1 | - | 1 | 1710 | 138 | |
Set Membership: |
|