Assay Details
| Gene Symbol: RGLG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106586.4 | NP_565206.1 | 8 - 9 | 1268 | 69 | |
| GenBank mRNA | AF372928.1 | - | 8 - 9 | 1268 | 69 | |
| AY133585.1 | - | 8 - 9 | 947 | 69 | ||
More Information
Set Membership: |
|

RGLG4
Genomic Map