Assay Details
| Gene Symbol: UBC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114513.6 | NP_566884.1 | 2 - 3 | 297 | 154 | |
| GenBank mRNA | AY050368.1 | - | 2 - 3 | 284 | 154 | |
| AY094040.1 | - | 2 - 3 | 147 | 154 | ||
| DQ027027.1 | - | 2 - 3 | 147 | 154 | ||
More Information
Set Membership: |
|

UBC13
Genomic Map