Assay Details
| Gene Symbol: SELT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114598.3 | NP_190314.2 | 1 - 2 | 210 | 86 | |
| GenBank mRNA | AK175455.1 | - | 1 - 2 | 179 | 86 | |
| AK176362.1 | - | 1 - 2 | 205 | 86 | ||
| BT010507.1 | - | 1 - 2 | 142 | 86 | ||
More Information
Set Membership: |
|

SELT
Genomic Map