Assay Details
| Gene Symbol: SELT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114598.3 | NP_190314.2 | 2 - 3 | 281 | 120 | |
| GenBank mRNA | AK175455.1 | - | 2 - 3 | 250 | 120 | |
| AK176362.1 | - | 2 - 3 | 276 | 120 | ||
| BT010507.1 | - | 2 - 3 | 213 | 120 | ||
More Information
Set Membership: |
|

SELT
Genomic Map