Assay Details
| Gene Symbol: RPN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118989.6 | NP_194576.5 | 14 - 15 | 1719 | 89 | |
| GenBank mRNA | AY230829.1 | - | 14 - 15 | 1553 | 89 | |
More Information
Set Membership: |
|

RPN1B
Genomic Map