Assay Details
| Gene Symbol: RPN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118989.6 | NP_194576.5 | 2 - 3 | 446 | 78 | |
| GenBank mRNA | AY230829.1 | - | 2 - 3 | 280 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPN1B| Gene Symbol: RPN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118989.6 | NP_194576.5 | 2 - 3 | 446 | 78 | |
| GenBank mRNA | AY230829.1 | - | 2 - 3 | 280 | 78 | |
Set Membership: |
|