Assay Details
| Gene Symbol: AT5G27850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344044.1 | NP_001332349.1 | 1 - 2 | 271 | 77 | |
| NM_122667.4 | NP_198137.1 | 2 - 3 | 190 | 77 | ||
| GenBank mRNA | AY056141.1 | - | 1 - 2 | 100 | 77 | |
| AY122996.1 | - | 1 - 2 | 88 | 77 | ||
| BT000711.1 | - | 1 - 2 | 100 | 77 | ||
| Gene Symbol: MLP34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230234.1 | - | 2198 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001344044.1 transcripts.
Set Membership: |
|

AT5G27850
Genomic Map