Assay Details
| Gene Symbol: CDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122858.3 | NP_198319.1 | 1 | 1099 | 76 | |
| GenBank mRNA | AY243479.1 | - | 1 | 1059 | 76 | |
| BT026129.1 | - | 1 | 1052 | 76 | ||
| DQ446998.1 | - | 1 | 1052 | 76 | ||
More Information
Set Membership: |
|

CDR1
Genomic Map