Assay Details
Gene Symbol: PTEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123302.4 | NP_198756.2 | 3 - 4 | 591 | 76 | |
GenBank mRNA | AJ490172.1 | - | 3 - 4 | 485 | 76 | |
AK118115.1 | - | 3 - 4 | 573 | 76 | ||
BT005560.1 | - | 2 - 3 | 455 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
