Assay Details
| Gene Symbol: CLPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123356.3 | NP_198809.2 | 4 - 5 | 680 | 86 | |
| GenBank mRNA | AY954872.1 | - | 4 - 5 | 581 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLPS5| Gene Symbol: CLPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123356.3 | NP_198809.2 | 4 - 5 | 680 | 86 | |
| GenBank mRNA | AY954872.1 | - | 4 - 5 | 581 | 86 | |
Set Membership: |
|