Assay Details
Gene Symbol: CLPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123356.3 | NP_198809.2 | 6 - 7 | 872 | 158 | |
GenBank mRNA | AY954872.1 | - | 6 - 7 | 773 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
