Assay Details
| Gene Symbol: ATSDI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124262.4 | NP_199696.2 | 1 - 2 | 290 | 75 | |
| GenBank mRNA | AK118038.1 | - | 1 - 2 | 196 | 75 | |
| BT005297.1 | - | 1 - 2 | 136 | 75 | ||
More Information
Set Membership: |
|

ATSDI1
Genomic Map