Assay Details
| Gene Symbol: PGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124561.3 | NP_199995.1 | 1 - 2 | 590 | 88 | |
| GenBank mRNA | AF216580.1 | - | 1 - 2 | 305 | 88 | |
| AY099708.1 | - | 1 - 2 | 261 | 88 | ||
| AY128901.1 | - | 1 - 2 | 196 | 88 | ||
More Information
Set Membership: |
|

PGM
Genomic Map