Assay Details
| Gene Symbol: BAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124585.4 | NP_200019.2 | 3 - 4 | 1319 | 84 | |
| GenBank mRNA | AK227095.1 | - | 3 - 4 | 1172 | 84 | |
More Information
Set Membership: |
|

BAG1
Genomic Map