Assay Details
Gene Symbol: OVA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124631.4 | NP_200065.1 | 1 - 2 | 418 | 84 | |
GenBank mRNA | AK176421.1 | - | 1 - 2 | 375 | 84 | |
BT010566.1 | - | 1 - 2 | 326 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
