Assay Details
| Gene Symbol: OVA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124631.4 | NP_200065.1 | 7 - 8 | 1132 | 83 | |
| GenBank mRNA | AK176421.1 | - | 7 - 8 | 1089 | 83 | |
| BT010566.1 | - | 7 - 8 | 1040 | 83 | ||
More Information
Set Membership: |
|

OVA6
Genomic Map