Assay Details
| Gene Symbol: SCAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129400.3 | NP_181378.2 | 6 - 7 | 4265 | 84 | |
| GenBank mRNA | AY743924.1 | - | 6 - 7 | 3950 | 84 | |
| AY817016.1 | - | 6 - 7 | 4171 | 84 | ||
More Information
Set Membership: |
|

SCAR2
Genomic Map