Assay Details
| Gene Symbol: SCAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129400.3 | NP_181378.2 | 7 - 8 | 4353 | 71 | |
| GenBank mRNA | AY743924.1 | - | 7 - 8 | 4038 | 71 | |
| AY817016.1 | - | 7 - 8 | 4259 | 71 | ||
More Information
Set Membership: |
|

SCAR2
Genomic Map