Assay Details
Gene Symbol: KT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084563.2 | NP_001078032.1 | 7 - 8 | 1717 | 101 | |
NM_001336844.1 | NP_001318392.1 | 8 - 9 | 1206 | 101 | ||
NM_001336845.1 | NP_001324081.1 | 8 - 9 | 1646 | 101 | ||
NM_001336846.1 | NP_001324082.1 | 8 - 9 | 1623 | 101 | ||
NM_129616.4 | NP_565936.1 | 8 - 9 | 1382 | 101 | ||
GenBank mRNA | AF012657.1 | - | 7 - 8 | 1007 | 101 | |
BT004049.1 | - | 8 - 9 | 1174 | 101 | ||
BT005048.1 | - | 7 - 8 | 1004 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
