Assay Details
| Gene Symbol: FUT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334647.1 | NP_001320576.1 | 1 - 2 | 421 | 65 | |
| NM_202413.2 | NP_974142.1 | 1 - 2 | 327 | 65 | ||
More Information
Set Membership: |
|

FUT3
Genomic Map