Assay Details
Gene Symbol: ACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036427.3 | NP_001031504.1 | 3 - 4 | 602 | 135 | |
NM_001336664.1 | NP_001324518.1 | 3 - 4 | 718 | 135 | ||
NM_001336665.1 | NP_001324519.1 | 3 - 4 | 714 | 135 | ||
NM_179953.3 | NP_850284.1 | 3 - 4 | 849 | 135 | ||
GenBank mRNA | AY074873.1 | - | 3 - 4 | 615 | 135 | |
AY093784.1 | - | 2 - 3 | 452 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
