Assay Details
Gene Symbol: ACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036427.3 | NP_001031504.1 | 4 - 5 | 1219 | 153 | |
NM_001336664.1 | NP_001324518.1 | 4 - 5 | 1335 | 153 | ||
NM_001336665.1 | NP_001324519.1 | 4 - 5 | 1331 | 153 | ||
NM_179953.3 | NP_850284.1 | 4 - 5 | 1466 | 153 | ||
GenBank mRNA | AY074873.1 | - | 4 - 5 | 1232 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
