Assay Details
| Gene Symbol: AGL53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122589.1 | NP_198059.1 | 1 | 647 | 125 | |
| GenBank mRNA | AY141217.1 | - | 1 | 647 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL53| Gene Symbol: AGL53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122589.1 | NP_198059.1 | 1 | 647 | 125 | |
| GenBank mRNA | AY141217.1 | - | 1 | 647 | 125 | |
Set Membership: |
|