Assay Details
| Gene Symbol: BRCC36A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124154.2 | NP_001117626.1 | 7 - 8 | 1094 | 119 | |
| NM_106670.4 | NP_178138.2 | 7 - 8 | 1094 | 119 | ||
| GenBank mRNA | AY093768.1 | - | 7 - 8 | 1071 | 119 | |
| BT002265.1 | - | 7 - 8 | 967 | 119 | ||
More Information
Set Membership: |
|

BRCC36A
Genomic Map