Assay Details
Gene Symbol: ALOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192792.2 | NP_001179721.1 | 5 - 6 | 754 | 80 | |
GenBank mRNA | AJ306424.1 | - | 5 - 6 | 664 | 80 | |
BC151634.1 | - | 5 - 6 | 755 | 80 | ||
EH163865.1 | - | 2 - 3 | 141 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
