Assay Details
| Gene Symbol: SESN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101174.1 | NP_001094644.1 | 9 - 10 | 1513 | 82 | |
| GenBank mRNA | BC102776.1 | - | 9 - 10 | 1514 | 82 | |
| BC149973.1 | - | 9 - 10 | 1513 | 82 | ||
| BC151575.1 | - | 9 - 10 | 1850 | 82 | ||
More Information
Set Membership: |
|

SESN1
Genomic Map