Assay Details
| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035076.2 | NP_001030248.1 | 2 - 3 | 314 | 131 | |
| GenBank mRNA | BC102302.1 | - | 2 - 3 | 317 | 131 | |
More Information
Set Membership: |
|

TMEM59
Genomic Map