Assay Details
| Gene Symbol: ARHGEF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035418.1 | NP_001030495.1 | 10 - 11 | 1528 | 76 | |
| GenBank mRNA | BT021580.1 | - | 10 - 11 | 1528 | 76 | |
More Information
Set Membership: |
|

ARHGEF9
Genomic Map