Assay Details
| Gene Symbol: PATZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191197.3 | NP_001178126.1 | 3 - 4 | 1503 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PATZ1| Gene Symbol: PATZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191197.3 | NP_001178126.1 | 3 - 4 | 1503 | 78 | |
Set Membership: |
|