Assay Details
| Gene Symbol: HNRNPDL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083725.2 | NP_001077194.1 | 1 - 2 | 710 | 60 | |
| GenBank mRNA | BC105386.1 | - | 1 - 2 | 189 | 60 | |
| BC134448.1 | - | 1 - 2 | 711 | 60 | ||
More Information
Set Membership: |
|

HNRNPDL
Genomic Map