Assay Details
| Gene Symbol: LYPLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080362.1 | NP_001073831.1 | 5 - 6 | 311 | 62 | |
| GenBank mRNA | BC118283.1 | - | 5 - 6 | 311 | 62 | |
| BT029895.1 | - | 5 - 6 | 328 | 62 | ||
More Information
Set Membership: |
|

LYPLA2
Genomic Map