Assay Details
| Gene Symbol: OTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177487.2 | NP_803453.1 | 8 - 9 | 925 | 85 | |
| GenBank mRNA | AF134841.1 | - | 8 - 9 | 925 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: OTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177487.2 | NP_803453.1 | 8 - 9 | 925 | 85 | |
| GenBank mRNA | AF134841.1 | - | 8 - 9 | 925 | 85 | |
Set Membership: |
|