Assay Details
| Gene Symbol: MTDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046038.1 | NP_001039503.1 | 3 - 4 | 584 | 71 | |
| GenBank mRNA | BC114169.1 | - | 3 - 4 | 584 | 71 | |
More Information
Set Membership: |
|

MTDH
Genomic Map