Assay Details
| Gene Symbol: WDR46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015564.1 | NP_001015564.1 | 10 - 11 | 1139 | 83 | |
| GenBank mRNA | BC134554.1 | - | 10 - 11 | 1155 | 83 | |
| BT020687.1 | - | 10 - 11 | 1139 | 83 | ||
More Information
Set Membership: |
|

WDR46
Genomic Map