Assay Details
| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173881.2 | NP_776306.1 | 1 - 2 | 168 | 103 | |
| GenBank mRNA | BC102986.1 | - | 1 - 2 | 170 | 103 | |
| D00409.1 | - | 162 | 103 | |||
| DT720521.1 | - | 1 - 2 | 169 | 103 | ||
More Information
Set Membership: |
|

MB
Genomic Map