Assay Details
| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173881.2 | NP_776306.1 | 2 - 3 | 384 | 98 | |
| GenBank mRNA | BC102986.1 | - | 2 - 3 | 386 | 98 | |
| D00409.1 | - | 378 | 98 | |||
| DT720521.1 | - | 2 - 3 | 385 | 98 | ||
More Information
Set Membership: |
|

MB
Genomic Map