Assay Details
| Gene Symbol: CA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173898.2 | NP_776323.1 | 1 - 2 | 134 | 117 | |
| GenBank mRNA | X96503.1 | - | 134 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_173898.2, X96503.1 transcripts.
Set Membership: |
|

CA6
Genomic Map